Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135823.1 | NP_001129295.1 | 3 | 503 | ||
NM_005086.4 | NP_005077.2 | 3 | 686 | |||
XM_011520853.2 | XP_011519155.1 | 3 | 436 | |||
XM_011520854.2 | XP_011519156.1 | 3 | 610 | |||
XM_011520855.2 | XP_011519157.1 | 3 | 352 | |||
GenBank mRNA | AF016028.2 | - | 3 | 586 | ||
AK092925.1 | - | 3 | 499 | |||
AK304667.1 | - | 4 | 580 | |||
AK314353.1 | - | 3 | 675 | |||
AL136756.1 | - | 3 | 670 | |||
BC062299.1 | - | 3 | 666 | 6376868 | ||
DA073350.1 | - | 4 | 471 | |||
X89105.1 | - | 3 | 508 |