Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134537.1 | NP_001128009.1 | 13 | 2012 | ||
XM_006249386.2 | XP_006249448.1 | 19 | 3097 | |||
GenBank mRNA | BC168650.1 | - | 19 | 3132 | 7121002 |