Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YEATS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006530.3 | NP_006521.1 | 3, 4 | 501 | ||
GenBank mRNA | AJ245746.1 | - | 3, 4 | 449 | ||
AK001413.1 | - | 3, 4 | 274 | |||
BC000994.2 | - | 3, 4 | 362 | 2899965 | ||
U61384.1 | - | 3, 4 | 459 |