Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YEATS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300950.1 | NP_001287879.1 | 5 | 716 | ||
NM_006530.3 | NP_006521.1 | 7 | 878 | |||
GenBank mRNA | AJ245746.1 | - | 7 | 826 | ||
AK001413.1 | - | 7 | 651 | |||
BC000994.2 | - | 7 | 739 | 2899965 | ||
U61384.1 | - | 7 | 836 |