Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YEATS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300950.1 | NP_001287879.1 | 1, 2 | 315 | ||
NM_006530.3 | NP_006521.1 | 1, 2 | 315 | |||
GenBank mRNA | AJ245746.1 | - | 1, 2 | 263 | ||
AK001413.1 | - | 1, 2 | 88 | |||
BC000994.2 | - | 1, 2 | 176 | 2899965 | ||
CA775118.1 | - | 1, 2 | 215 | |||
U61384.1 | - | 1, 2 | 273 |