Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280542.1 | NP_001267471.1 | 4, 5 | 460 | ||
NM_001280543.1 | NP_001267472.1 | 5, 6 | 604 | |||
NM_001280544.1 | NP_001267473.1 | 4, 5 | 903 | |||
NM_012074.4 | NP_036206.3 | 4, 5 | 460 | |||
XM_017021670.1 | XP_016877159.1 | 4, 5 | 460 | |||
XM_017021671.1 | XP_016877160.1 | 4, 5 | 1210 | |||
XM_017021672.1 | XP_016877161.1 | 4, 5 | 1212 | |||
XM_017021673.1 | XP_016877162.1 | 4, 5 | 800 | |||
XM_017021674.1 | XP_016877163.1 | 4, 5 | 989 | |||
XM_017021675.1 | XP_016877164.1 | 4, 5 | 1213 | |||
GenBank mRNA | AK024141.1 | - | 4, 5 | 445 | ||
AK126933.1 | - | 4, 5 | 903 | |||
AK294425.1 | - | 5, 6 | 589 | |||
AY803021.1 | - | 4, 5 | 432 | |||
BC026305.1 | - | 4, 5 | 445 | |||
BC060801.1 | - | 4, 5 | 445 | 5260159 | ||
DA010060.1 | - | 4, 5 | 446 | |||
DA407279.1 | - | 4, 5 | 482 | |||
DB140241.1 | - | 4, 5 | 460 |