Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Optc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107176.1 | NP_001100646.1 | 2 | 483 | ||
XM_006249871.2 | XP_006249933.1 | 3 | 1330 | |||
XM_006249872.2 | XP_006249934.1 | 3 | 1889 | |||
XM_008769554.1 | XP_008767776.1 | 2 | 1716 |