Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024768.1 | NP_001019939.1 | 8 | 1135 | ||
XM_006249881.2 | XP_006249943.1 | 8 | 1230 | |||
XM_006249883.2 | XP_006249945.1 | 8 | 1161 | |||
GenBank mRNA | BC097980.1 | - | 8 | 1135 | 7443278 |