Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107179.1 | NP_001100649.1 | 2 | 1589 | ||
XM_008769562.1 | XP_008767784.1 | 2 | 2017 |