Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177676.1 | NP_001171147.1 | 2 | 365 | ||
NM_003485.3 | NP_003476.3 | 2 | 319 | |||
XM_005268110.4 | XP_005268167.1 | 2 | 1505 | |||
XM_005268111.3 | XP_005268168.1 | 2 | 285 | |||
XM_005268112.3 | XP_005268169.1 | 2 | 225 | |||
XM_006720262.3 | XP_006720325.1 | 2 | 251 | |||
XM_011537196.2 | XP_011535498.1 | 3 | 369 | |||
XM_011537197.2 | XP_011535499.1 | 2 | 649 | |||
XM_011537198.2 | XP_011535500.1 | 2 | 875 | |||
XM_011537199.2 | XP_011535501.1 | 2 | 320 | |||
GenBank mRNA | AY615372.1 | - | 1 | 54 | ||
BC067470.1 | - | 1 | 24 | |||
BC067471.1 | - | 1 | 24 | |||
BC067472.1 | - | 1 | 24 | |||
BC069592.1 | - | 1 | 24 | 7262355 | ||
BC096071.3 | - | 1 | 90 | 40000421 | ||
BC096072.3 | - | 1 | 90 | 40000422 | ||
BC096073.1 | - | 1 | 90 | 40000423 | ||
BC096074.3 | - | 1 | 90 | 40000425 | ||
BC098567.1 | - | 2 | 262 | 30925589 | ||
BC128536.1 | - | 2 | 373 | 40120393 | ||
BM723705.1 | - | 2 | 364 | |||
EU883575.1 | - | 1 | 54 | |||
U35398.1 | - | 1 | 124 |
Set Membership: |
G-protein coupled receptor |