Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANP32A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006305.3 | NP_006296.1 | 2 | 346 | ||
GenBank mRNA | AF025684.1 | - | 2 | 323 | ||
AK127498.1 | - | 2 | 3131 | |||
AK223280.1 | - | 2 | 292 | |||
AK312703.1 | - | 2 | 319 | |||
AY349171.1 | - | 2 | 175 | |||
BC000608.2 | - | 2 | 310 | |||
BC007200.1 | - | 2 | 308 | 3677623 | ||
BC106719.2 | - | 2 | 175 | 40014106 | ||
BC125143.1 | - | 2 | 179 | 40119966 | ||
BT007436.1 | - | 2 | 175 | |||
DA825897.1 | - | 2 | 346 | |||
U60823.1 | - | 2 | 175 | |||
U73477.1 | - | 2 | 271 | |||
X75090.1 | - | 2 | 278 |