Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107197.1 | NP_001100667.1 | 5 | 740 | ||
XM_006250290.2 | XP_006250352.1 | 5 | 917 | |||
XM_006250294.2 | XP_006250356.1 | 5 | 918 | |||
XM_008769740.1 | XP_008767962.1 | 5 | 920 | |||
XM_008769741.1 | XP_008767963.1 | 5 | 1135 | |||
XM_008769742.1 | XP_008767964.1 | 5 | 752 | |||
XM_008769743.1 | XP_008767965.1 | 5 | 650 |