Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107197.1 | NP_001100667.1 | 7 | 1073 | ||
XM_006250290.2 | XP_006250352.1 | 7 | 1250 | |||
XM_006250294.2 | XP_006250356.1 | 7 | 1251 | |||
XM_008769740.1 | XP_008767962.1 | 7 | 1253 | |||
XM_008769741.1 | XP_008767963.1 | 7 | 1468 | |||
XM_008769742.1 | XP_008767964.1 | 7 | 1085 | |||
XM_008769743.1 | XP_008767965.1 | 7 | 983 |