Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250335.1 | XP_006250397.1 | 2 | 365 | ||
XM_006250336.2 | XP_006250398.1 | 3 | 809 | |||
XM_006250339.1 | XP_006250401.1 | 1 | 180 | |||
XM_008769802.1 | XP_008768024.1 | 3 | 2024 | |||
XM_008769803.1 | XP_008768025.1 | 3 | 2974 | |||
XM_008769804.1 | XP_008768026.1 | 4 | 2352 | |||
XM_008769805.1 | XP_008768027.1 | 3 | 1961 | |||
XM_222949.8 | XP_222949.5 | 2 | 2789 |