Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sccpdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013985.1 | NP_001014007.1 | 2, 3 | 414 | ||
GenBank mRNA | BC061579.1 | - | 348 | |||
BC079215.1 | - | 2, 3 | 414 | 7131885 |
Gene Symbol: LOC103689999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769879.1 | XP_008768101.1 | 2, 3 | 596 |