Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001663.3 | NP_001654.1 | 2 | 877 | ||
GenBank mRNA | AF047432.1 | - | 1 | 323 | ||
AF493885.1 | - | 1 | 256 | |||
AK313790.1 | - | 2 | 796 | |||
BC002952.1 | - | 2 | 780 | |||
BC008918.2 | - | 2 | 792 | 3626100 | ||
BT019868.1 | - | 1 | 256 | |||
CR541939.1 | - | 1 | 256 | |||
CR541964.1 | - | 1 | 256 | |||
M57763.1 | - | 2 | 773 |