Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107201.1 | NP_001100671.1 | 4 | 2476 | ||
XM_006250454.2 | XP_006250516.1 | 4 | 2606 | |||
XM_006250455.2 | XP_006250517.1 | 4 | 2303 | |||
XM_006250456.2 | XP_006250518.1 | 4 | 2925 | |||
XM_006250458.2 | XP_006250520.1 | 4 | 2606 |