Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013987.1 | NP_001014009.1 | 5 | 386 | ||
XM_006250547.2 | XP_006250609.1 | 5 | 511 | |||
XM_006250548.2 | XP_006250610.2 | 5 | 501 | |||
XM_008769928.1 | XP_008768150.1 | 5 | 501 | |||
XM_008769929.1 | XP_008768151.1 | 5 | 501 | |||
XM_008769930.1 | XP_008768152.1 | 3 | 172 | |||
GenBank mRNA | BC088426.1 | - | 5 | 386 | 7123983 |