Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100564.1 | NP_001094034.1 | 3 | 882 | ||
XM_006250657.2 | XP_006250719.1 | 4 | 998 | |||
XM_006250658.2 | XP_006250720.1 | 4 | 1028 | |||
XM_006250659.2 | XP_006250721.1 | 4 | 1193 | |||
XM_006250661.2 | XP_006250723.1 | 4 | 365 | |||
XM_008770022.1 | XP_008768244.1 | 4 | 335 | |||
GenBank mRNA | BC082043.1 | - | 1 | 88 | ||
EV774847.1 | - | 3 | 489 |