Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100890.1 | NP_001094360.1 | 2 | 461 | ||
XM_006250775.2 | XP_006250837.1 | 2 | 348 | |||
GenBank mRNA | FM050032.1 | - | 2 | 422 |