Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004098.1 | NP_001004098.1 | 10 | 1644 | ||
GenBank mRNA | AY693786.1 | - | 3 | 487 | ||
AY693787.1 | - | 5 | 1068 | |||
AY693788.1 | - | 10 | 1644 |