Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slain2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107214.1 | NP_001100684.1 | 4 | 1002 | ||
XM_006250898.2 | XP_006250960.1 | 4 | 1074 | |||
XM_006250899.2 | XP_006250961.1 | 4 | 1073 | |||
GenBank mRNA | BC158815.1 | - | 2 | 75 |