Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107215.1 | NP_001100685.1 | 9, 10 | 1198 | ||
XM_008770207.1 | XP_008768429.1 | 9, 10 | 1601 |