Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146035.1 | NP_001139507.1 | 1 | 1547 | ||
XM_006251002.2 | XP_006251064.1 | 2 | 1653 | |||
GenBank mRNA | FJ755908.1 | - | 1 | 1547 |