Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173341.1 | NP_001166812.1 | 5 | 553 | ||
XM_006251014.2 | XP_006251076.1 | 5 | 641 | |||
GenBank mRNA | BC158739.1 | - | 4 | 493 | 9099241 | |
EV772004.1 | - | 5 | 574 |
Gene Symbol: LOC100912369 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751682.3 | XP_003751730.1 | 5 | 641 | ||
XM_006253886.2 | XP_006253948.1 | 5 | 582 |