Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007165.4 | NP_009096.2 | 7 | 576 | ||
GenBank mRNA | AK314815.1 | - | 7 | 569 | ||
BC004434.1 | - | 7 | 546 | 2820900 | ||
BC009903.2 | - | 7 | 569 | 2959124 | ||
BC015804.1 | - | 7 | 529 |