Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Urgcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077660.2 | NP_001071128.1 | 6 | 2401 | ||
XM_006251312.2 | XP_006251374.1 | 6 | 2862 | |||
XM_006251313.2 | XP_006251375.1 | 6 | 2691 | |||
XM_006251315.2 | XP_006251377.1 | 6 | 2494 | |||
XM_008770307.1 | XP_008768529.1 | 5 | 2383 | |||
GenBank mRNA | EX490902.1 | - | 1 | 467 | ||
FQ222248.1 | - | 6 | 2393 | |||
FQ223767.1 | - | 6 | 2388 |