Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107237.1 | NP_001100707.1 | 4, 5 | 544 | ||
XM_006251491.2 | XP_006251553.1 | 5, 6 | 1317 | |||
XM_006251492.2 | XP_006251554.1 | 5, 6 | 5152 | |||
XM_006251493.2 | XP_006251555.1 | 5, 6 | 855 | |||
XM_006251494.2 | XP_006251556.2 | 5, 6 | 1142 | |||
XM_006251495.2 | XP_006251557.1 | 5, 6 | 830 | |||
XM_006251496.2 | XP_006251558.1 | 5, 6 | 629 | |||
XM_006251497.2 | XP_006251559.1 | 5, 6 | 863 | |||
XM_008770312.1 | XP_008768534.1 | 5, 6 | 1142 | |||
XM_008770314.1 | XP_008768536.1 | 5, 6 | 1142 | |||
XM_008770315.1 | XP_008768537.1 | 4, 5 | 884 | |||
XM_008770317.1 | XP_008768539.1 | 4, 5 | 884 |