Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107239.1 | NP_001100709.1 | 8 | 1394 | ||
XM_008770320.1 | XP_008768542.1 | 10 | 1624 | |||
XM_008770321.1 | XP_008768543.1 | 9 | 1552 | |||
XM_008770322.1 | XP_008768544.1 | 8 | 973 |