Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107239.1 | NP_001100709.1 | 17 | 2624 | ||
XM_008770320.1 | XP_008768542.1 | 19 | 2854 | |||
XM_008770321.1 | XP_008768543.1 | 18 | 2782 | |||
XM_008770322.1 | XP_008768544.1 | 17 | 2203 |