Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdpcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127537.1 | NP_001121009.1 | 10 | 1228 | ||
GenBank mRNA | BC161962.1 | - | 10 | 1228 | 9027983 |