Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135155.2 | NP_001128627.1 | 3 | 316 | ||
NM_001135156.2 | NP_001128628.1 | 3 | 330 | |||
NM_001289978.1 | NP_001276907.1 | 3 | 316 | |||
NM_004647.3 | NP_004638.2 | 3 | 316 | |||
XM_005259289.2 | XP_005259346.1 | 3 | 316 | |||
XM_005259290.3 | XP_005259347.1 | 3 | 287 | |||
XM_005259292.4 | XP_005259349.1 | 3 | 259 | |||
XM_006723407.2 | XP_006723470.1 | 3 | 287 | |||
XM_006723408.3 | XP_006723471.1 | 3 | 261 | |||
XM_006723409.2 | XP_006723472.1 | 3 | 316 | |||
XM_011527356.1 | XP_011525658.1 | 3 | 316 | |||
XM_011527357.2 | XP_011525659.1 | 3 | 262 | |||
XM_011527358.2 | XP_011525660.1 | 3 | 331 | |||
GenBank mRNA | AK094632.1 | - | 3 | 643 | ||
AK297581.1 | - | 4 | 290 | |||
AK308563.1 | - | 3 | 316 | |||
AL529673.3 | - | 3 | 298 | |||
AL538568.3 | - | 3 | 335 | |||
BC021191.2 | - | 3 | 289 | |||
BC125152.1 | - | 3 | 218 | |||
BC125153.1 | - | 3 | 218 | 40120602 | ||
GQ358728.1 | - | 3 | 208 | |||
U43843.1 | - | 3 | 214 |