Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000557.4 | NP_000548.2 | 2 | 1718 | ||
NM_001319138.1 | NP_001306067.1 | 4 | 1960 | |||
XM_011529075.2 | XP_011527377.1 | 4 | 1850 | |||
GenBank mRNA | AB593125.1 | - | 2 | 1706 | ||
BC032495.1 | - | 4 | 1960 | 5243109 | ||
BC111566.1 | - | 1 | 625 | |||
BC111859.1 | - | 1 | 625 | |||
U13660.1 | - | 2 | 1718 |