Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fezf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107251.1 | NP_001100721.1 | 5 | 1569 | ||
XM_006251737.2 | XP_006251799.1 | 7 | 1832 | |||
GenBank mRNA | BC168214.1 | - | 5 | 1399 | 7313369 |