Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033066.1 | NP_001028238.1 | 10 | 2100 | ||
XM_006251769.2 | XP_006251831.1 | 12 | 2291 | |||
XM_006251770.2 | XP_006251832.1 | 11 | 2270 | |||
XM_006251771.2 | XP_006251833.1 | 12 | 2291 | |||
XM_006251772.2 | XP_006251834.1 | 11 | 2189 | |||
XM_006251773.2 | XP_006251835.1 | 10 | 2168 | |||
XM_006251774.2 | XP_006251836.1 | 11 | 2188 | |||
GenBank mRNA | BC103649.1 | - | 10 | 2100 | 7314879 |