Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007681.1 | NP_001007682.1 | 2 | 542 | ||
XM_006251878.2 | XP_006251940.1 | 3 | 1701 | |||
XM_006251879.2 | XP_006251941.1 | 4 | 1359 | |||
XM_006251880.2 | XP_006251942.1 | 3 | 1085 | |||
XM_006251882.2 | XP_006251944.1 | 2 | 996 | |||
XM_008770522.1 | XP_008768744.1 | 3 | 1385 | |||
GenBank mRNA | BC082017.1 | - | 2 | 542 | 7132141 | |
FQ212172.1 | - | 3 | 525 | |||
FQ221971.1 | - | 4 | 949 |