Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033257.3 | NP_150282.2 | 5 | 1147 | ||
GenBank mRNA | AF228708.1 | - | 5 | 1127 | ||
AK001528.1 | - | 5 | 1088 | |||
AK289952.1 | - | 5 | 1065 | |||
AK307210.1 | - | 4 | 982 | |||
BC000682.2 | - | 5 | 1083 | 3349309 |
Gene Symbol: DGCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005675.4 | NP_005666.2 | 5 | 1182 | ||
XM_011530394.1 | XP_011528696.1 | 6 | 1169 | |||
GenBank mRNA | AF228707.1 | - | 5 | 1126 | ||
AK098780.1 | - | 5 | 1404 | |||
BC047039.1 | - | 5 | 1075 | 5229172 | ||
X96484.1 | - | 5 | 1044 |
Gene Symbol: LOC102724770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724934.1 | XP_006724997.1 | 5 | 1124 |