Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107265.1 | NP_001100735.1 | 8 | 2899 | ||
XM_008770747.1 | XP_008768969.1 | 9 | 6337 |