Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006767.3 | NP_006758.2 | 5 | 552 | ||
GenBank mRNA | AK296347.1 | - | 4 | 433 | ||
AK310747.1 | - | 3 | 155 | |||
BC026214.1 | - | 5 | 454 | 4399716 | ||
CT841521.1 | - | 5 | 468 | |||
D38496.1 | - | 5 | 446 |