Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191686.1 | NP_001178615.1 | 9, 10 | 3238 | ||
XM_006252113.2 | XP_006252175.1 | 9, 10 | 3305 | |||
XM_006252114.2 | XP_006252176.1 | 8, 9 | 3779 | |||
XM_006252115.2 | XP_006252177.1 | 6, 7 | 2279 |