Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx3-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034144.1 | NP_001029316.1 | 2 | 501 | ||
GenBank mRNA | BC100628.1 | - | 2 | 501 | 7938032 |