Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012227.3 | NP_036359.3 | 8 | 1278 | ||
XM_006724868.3 | XP_006724931.1 | 9 | 1178 | |||
XM_011545637.2 | XP_011543939.1 | 7 | 883 | |||
GenBank mRNA | AK024630.1 | - | 8 | 1217 | ||
AK026462.1 | - | 2 | 299 | |||
AK074528.1 | - | 2 | 1166 | |||
AK097872.1 | - | 3 | 1616 | |||
AK223412.1 | - | 8 | 1223 | |||
BC014636.1 | - | 8 | 951 | 3528598 | ||
BT007360.1 | - | 7 | 907 | |||
Y14391.2 | - | 8 | 1236 |