Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pibf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252404.2 | XP_006252466.1 | 12 | 1853 | ||
XM_006252405.2 | XP_006252467.1 | 12 | 1806 | |||
XM_006252406.2 | XP_006252468.1 | 12 | 1638 | |||
XM_006252407.2 | XP_006252469.1 | 12 | 1853 | |||
XM_224451.8 | XP_224451.3 | 12 | 1679 | |||
GenBank mRNA | FQ210517.1 | - | 1841 |