Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100568.1 | NP_001094038.1 | 5 | 903 | ||
XM_006252442.2 | XP_006252504.1 | 5 | 1120 | |||
GenBank mRNA | BC092649.1 | - | 3 | 415 |