Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252478.2 | XP_006252540.1 | 5, 6 | 550 | ||
XM_224534.8 | XP_224534.7 | 6, 7 | 796 |