Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182821.1 | NP_877973.1 | 1, 2 | 191 | ||
XM_006251775.1 | XP_006251837.1 | 1, 2 | 219 | |||
XM_006251776.2 | XP_006251838.1 | 1, 2 | 219 | |||
GenBank mRNA | AY327408.1 | - | 1, 2 | 191 | ||
BC098901.1 | - | 1, 2 | 133 | 7127537 |