Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771024.1 | XP_008769246.1 | 14 | 2008 | ||
XM_008771025.1 | XP_008769247.1 | 14 | 1963 | |||
XM_008771026.1 | XP_008769248.1 | 15 | 2134 | |||
XM_008771027.1 | XP_008769249.1 | 14 | 2108 | |||
XM_008771028.1 | XP_008769250.1 | 13 | 1918 | |||
XM_008771029.1 | XP_008769251.1 | 14 | 2050 |