Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nisch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001057307.4 | XP_001057307.4 | 21 | 4561 | ||
GenBank mRNA | BC098837.1 | - | 2 | 84 |