Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181632.2 | NP_853663.2 | 2, 3 | 316 | ||
GenBank mRNA | AB109021.1 | - | 1, 2 | 211 | ||
BC084685.1 | - | 2, 3 | 316 | 7100793 |