Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044230.2 | NP_001037695.1 | 8 | 2481 | ||
GenBank mRNA | DQ479969.1 | - | 8 | 2459 |
Gene Symbol: Gdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044240.2 | NP_001037705.1 | 8 | 2481 | ||
GenBank mRNA | DQ481538.1 | - | 4 | 1599 |